Learn More
Electron Microscopy Sciences Globe Slides One-Sided Frosted, Ground Edge, 90° Corner
SDP
Supplier:  Electron Microscopy Sciences 7188320
Encompass_Preferred
These frosted microscope slides are produced using a unique chemical etching process. This provides the slides with a smooth frosted finish that won't wear out slide marking pens.
Edge: Ground
Corner: 90°
Catalog No. 50-192-8787