Learn More
Newcomer Supply Slide Moisture Chamber, Black. 10 slide chamber for keeping your slides well separated, maintaining moisture, and watching the reaction. Stackable, heavy duty plastic construction.
SDP
Supplier:  Newcomer Supply 68432A
Encompass
The Slide Staining Tray / Moisture Chamber is perfect for keeping your slides well separated and maintaining moisture. The tray is divided into 10 individual compartments with 1/2" empty space between compartments. Eight barrier dividers are placed into empty spaces between compartments when lid is closed for complete isolation. Slides are placed on 4 pedestal posts to raise the surface above the water below. The trays are stackable and made of heavy duty polystyrene. The black slide staining tray chamber is effective for for light sensitive chemistry. Dimensions: 8 1/4" x 7" x 1 1/4". Temperature Rating: -10°C to 70°C. Clear Slide Staining Tray / Moisture Chamber also available to allow easy visualization of the reaction.
Catalog No. NC9062083