Learn More
Research Products International Corp Slide Tray for Immuno-Staining with Clear Cover
SDP
Supplier:  Research Products International Corp 195800
Encompass
Ideal for immunostaining and other applications that requires a humid chambers for processing. Allows safe and convenient slide processing using only one hand. The black tray base is molded from durable ABS plastic that provides excellent chemical resistance. Holds up to twenty slides on four plastic rails. A thin polymer strip on each rail holds slides level. Deep wells between rails are designed to hold water securely without splashing for procedures requiring humidity. A built-in drain plug allows a convenient emptying of contents. The clear cover allows for visual examination. Made of PETG with a temperature range of -20 °C to +60 °C.
Catalog No. 50-998-210