Learn More
Med Vet International Heathrow Hs15994A, 100-Place Slide Box, Blue, Cork, Each
Supplier:  Med Vet International HS15994A
Encompass_Preferred
Slide boxes are perfect for storage or transportation of standard size slides (25 x 75 mm and 1 x 3")Molded, rigid boxes securely hold slides in grooved slots. Keep boxes safely on the benchtop during use or stack boxes for long-term storage Both the lining and the slots are numbered to correspond to the slide inventory sheet on the inside cover of the box Product description Slide boxes are perfect for storage or transportation of standard size slides (25 x 75 mm and 1 x 3"). Molded, rigid boxes securely hold slides in grooved slots. Keep boxes safely on the benchtop during use or stack boxes for long-term storage. Both the lining and the slots are numbered to correspond to the slide inventory sheet on the inside cover of the box.
Catalog No. 50-283-2271