Learn More
SLINGSHOT BIOSCIENCES Slingshot Biosciences CD34+ Stem Cell Mimic Controls, Lyophilized, CD34/CD45/Viability, Immunophenotyping
SDP
Supplier:  SLINGSHOT BIOSCIENCES SL00046
Encompass_Preferred
CD34+ Stem Cell Mimic™ is a lyophilized cell mimic for immunophenotyping and flow cytometry. It features CD34, CD45, and DNA-based viability biomarkers, with scatter characteristics resembling human lymphocyte populations. A high density CD34+ population (∼90% of CD45+ cells) allows confident discrimination and gating, comparable to biological samples.
  • Lyophilized cell mimic; includes CD45+, CD45+/CD34+, and CD45+ with viability populations.
  • ∼90% CD34+ frequency (of CD45+ cells).
  • Supports CD34 enumeration, viability, antibody qualification, and staining verification.
  • Compatible across diverse cytometry platforms for multi-site standardization.
  • No special handling or safety precautions needed.
  • Extended shelf life for long-term assay harmonization.
Catalog No. NC3981035