Learn More
Aviva Systems Biology Corp SLITRK1 Antibody - N-terminal region (ARP50218_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50218_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLITRK1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLITRK1. Alternate names: KIAA0918, KIAA1910, LRRC12, RP11-395N17.1, TTMApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60289