Learn More
Medchemexpress LLC SLITRK4 Human HEK2 10ug | 10ug
SDP
Supplier:  Medchemexpress LLC HYP7720710UG
Encompass_Preferred
SLITRK4 Protein is pivotal in synaptogenesis promoting synapse differentiation and synaptic maturation Conversely it suppresses neurite outgrowth indicating a regulatory role in neuronal morphology Interacting with PTPRD through its LRR 1 and 2 repeats SLITRK4 suggests a potential role in modulating the activity or function of PTPRD a receptor-type protein tyrosine phosphatase associated with neural development and synaptic plasticity SLITRK4 Protein Human (HEK293 His) is the recombinant human-derived SLITRK4 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-86429