Learn More
Medchemexpress LLC HY-P0301A 1mg Medchemexpress, SLLK, Control Peptide for TSP1 Inhibitor(TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0301A1MG
Encompass_Preferred
Medchemexpress, HY-P0301A 1mg SLLK, Control Peptide for TSP1 Inhibitor(TFA) CAS: SLLK, Control Peptide for TSP1 Inhibitor (TFA) is a control peptide for LSKL , which is a Thrombospondin (TSP-1) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4107