Learn More
Sciencix Solvent Filter Adapter ETFE with 1/4-28 Female Port for use with CTS-10843
SDP
Supplier:  Sciencix 111888
Encompass
Solvent Filter Adapter ETFE with 1/4-28 Female Port for use with CTS-10843 (adapts glass filter for use with Mobile Phase Tubing 1mm to 1/8" OD)
Catalog No. NC2897387