Learn More
MSC Daubert Silver Saver, 18 in x 200 yd, 1 Case
Supplier:  MSC 39675566
Encompass
Silver Saver anti-tarnish paper is a 40 lb/ream (65 g/m²) bleached kraft coated with a proprietary chemical formulation that prevents silver and palladium from tarnishing. It protects silver and palladium without leaving residue on metal surfaces and does not affect solder, conductivity, or surface resistivity.
  • Prevents silver and palladium from tarnishing
  • Absorbs hydrogen sulfide and other sulfurous gases
  • Non-toxic, safe, and RoHS2 compliant
  • OEM approved and sulfur-free
  • Protects for up to 24 months
  • Does not affect solder, conductivity, or surface resistivity
  • Constructed from 40 lb/ream bleached kraft
Catalog No. 50-191-9842