Learn More
Apexbio Technology LLC SLx-2119, 911417-87-3, MFCD23098791, 10 mg
SDP
Supplier:  Apexbio Technology LLC A382510
Encompass_Preferred
MF: C26H24N6O2, MW: 452.51, Solubility: 22.65 mg/mL in DMSO, 26.4 mg/mL in EtOH with ultrasonic and warming,insoluble in H2O. SLx-2119(KD-025) is a selective inhibitor of ROCK2 with IC50 of 105 nM.
Catalog No. 50-101-4909