Learn More
Apexbio Technology LLC SLx-2119 911417-87-3 5mg
SDP
Supplier:  Apexbio Technology LLC A38255
Encompass_Preferred
SLx-2119 (CAS 911417-87-3) also known as KD-025 is a selective inhibitor targeting Rho-associated protein kinase 2 (ROCK2) exhibiting an IC50 of approximately 105 nM ROCK2 is a serine/threonine kinase central to cytoskeletal modulation and cellular signaling particularly expressed within neuronal and vascular cells SLx-2119 reduces connective tissue growth factor (CTGF) mRNA expression selectively in human intestinal smooth muscle cells isolated from radiation-induced fibrosis tissue (RE-SMC) whereas normal smooth muscle cells remain unaffected Additionally in a murine model of focal cerebral ischemia SLx-2119 treatment yields a dose-dependent decrease in infarct size across aged and diabetic models highlighting its research applicability in ischemic stroke and fibrotic disorders
Catalog No. 50-000-64346