Learn More
Medchemexpress LLC Slx-4090 | 913541-47-6 | MFCD28502203 | 99.0% | 546.54 | C31H25F3N2O4 | 10 MG

Supplier: Medchemexpress LLC HY10601110MG
SLX-4090 is an orally active, enterocytic-specific microsomal triglyceride transfer protein (MTP) inhibitor intended for research use. It exhibits potent MTP inhibition and is supplied with high purity in both solid and solution formats for in vitro and dyslipidemia research.
- Orally active, enterocytic-specific microsomal triglyceride transfer protein inhibitor.
- Potent in vitro activity (IC50 8.0 nM against MTP).
- Suitable for dyslipidemia research.
- High purity (~99%).
- Available as solids (5-500 mg) and 10 mM solution in DMSO.
- Molecular weight 546.54 g/mol.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.