Learn More
Medchemexpress LLC HY-134541 100mg , SM-102 CAS:2089251-47-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY134541100MG
Encompass_Preferred
HY-134541 100mg SM-102 CAS: 2089251-47-6 SM-102 is an ionizable amino lipid that can be used for the formation of lipid nanoparticles (LNPs). SM-102 has the potential for development of lipid nanoparticles for delivery of mRNA-based vaccines. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9138