Learn More
Medchemexpress LLC SM-7368 10mg | 380623-76-7 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11662610MG
Encompass_Preferred
SM-7368 is a small-molecule NF-κB inhibitor used for in vitro research. It targets downstream MAPK p38 activation and suppresses TNF-α-induced MMP-9 upregulation; intended for research use only.
  • Small-molecule NF-κB inhibitor targeting MAPK p38 downstream.
  • Reduces TNF-α-induced MMP-9 expression in cellular models.
  • Provided as a solid powder suitable for solution preparation in assays.
  • Available in small pack sizes for screening and cell-based experiments.
  • High reported purity and defined molecular properties for reproducible results.
Catalog No. 50-002-27053