Learn More
Medchemexpress LLC SM-7368 25mg | 380623-76-7 | 25 MG

Supplier: Medchemexpress LLC HY11662625MG
SM-7368 is a small-molecule NF-κB inhibitor for research use that inhibits TNF-α-induced MMP-9 upregulation and acts downstream of MAPK p38, making it suitable for studies of TNF-α-mediated tumor invasion and metastasis.
- High purity: 99.77% (HPLC).
- Molecular weight: 328.69 g·mol⁻¹.
- Chemical formula: C10H5ClN4O5S.
- Available in research-scale solid and solution pack sizes, including 25 MG solid and 10 mM in DMSO.
- Includes structural identifiers (SMILES) for computational and cheminformatics use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.