Learn More
HARVARD BIOSCIENCE INC SM DMTER RND CVR GLASS 100/PK
Supplier:  HARVARD BIOSCIENCE INC 640720
Encompass_Preferred
Total order minimum requirement of $500. If only item on order, must order QTY of 4.High quality coverslip glass is essential for microscopy and imaging. For microscopy at higher resolutions (e.g., 40X, 60X or 100X), choosing the correct coverslip thickness is critical to good image resolution.
Catalog No. NC0026170