Learn More
Sigma Aldrich Fine Chemicals Biosciences Sma I from Serratia marces
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 10656348001
Encompass
Sma I recognizes the sequence *CC*C8595GGG and generates fragments with blunt ends.
Catalog No. 50-290-4511