Learn More
Enzo Life Sciences Smac DIABLO monoclonal antibody (79-1-83) (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905684100
Encompass_Preferred
Clone: 79-1-83. Host: Mouse. Isotype: IgG2b. Immunogen: Recombinant full length human Smac/DIABLO. Source: Purified from mouse ascites. Species reactivity: Human, Monkey. Applications: IHC (PS), WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide and 50% glycerol.
Catalog No. 50-200-6992