Learn More
Sino Biological Recombinant Human NFAT2/NFATC1 Protein (Full Length, GST Tag)
Supplier:  Sino Biological N1230G100
Encompass_Preferred
Recombinant full length human NFATC1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4057