Learn More
Medchemexpress LLC Small interfering RNA (siRNA) negative control | 99.2% | 13323.03 Da | 10 MG

Supplier: Medchemexpress LLC HY15015010MG
SiRNA negative control is a 21-nucleotide non-targeting small interfering RNA used as a negative control in RNA interference experiments. It is supplied in lyophilized form, characterized for purity and stability, and intended for use in cell-based gene-silencing assays and transfection controls.
- Acts as a non-targeting control for RNAi experiments.
- 21-nucleotide length suitable for standard siRNA transfections.
- High purity (99.24%) to support consistent experimental results.
- Available in multiple package sizes, including a 10 mg option.
- Storage: -20°C sealed; in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month.
- Molecular weight approximately 13,323.03 Da.
- Intended for use in in vitro cell-based assays as a negative control.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.