Learn More
Electron Microscopy Sciences High Opacity Paint Markers Small Red
SDP
Supplier:  Electron Microscopy Sciences 72167R1
Encompass_Preferred
Constructed aluminum barrel, actuating ball and hard felt tips, these markers permanently cover nearly any surface.Weatherproof. Resistant to alcohol and acetone. Contain xylene.
Catalog No. 50-949-073