Learn More
Grainger Small Parts Drawer Unit 30 Drawers Clr
Supplier:  Grainger 157H12
Encompass_Preferred
The Treston ESD Small Parts Drawer Unit is a storage cabinet with 12 drawers designed for storing small parts and components. It is made of steel with a powder-coated finish and measures 21"W x 6"D x 12"H. The drawers are 3"H x 16.5"W x 5.5"D each and feature full extension slides for easy access. This ESD cabinet has a dissipative laminate worksurface and is grounded with a 10' cord and wrist strap. The unit can hold up to 55 lbs per drawer. It provides secure, organized storage for small parts and protects static sensitive items. This industrial drawer cabinet comes fully assembled and is made in Finland.
Catalog No. 50-275-2740