Learn More
Brookfield Engineer Labs SMALL SAMPLE ADAPT/WSPND/CHMBR
Supplier:  Brookfield Engineer Labs SSA29/13R
Encompass
Small Sample Adapterincludes Water Jacket, Mounting Bracket with Hardware, SC4-29 Spindle, SC4 -13R Sample Chamber, Storage Case
Catalog No. NC1513226