Learn More
LabScientific Small Slide Warmer with Temperature Control LED 110V; 120V (USA and Canada), Surface holds about 23 slides. Heats up to 75 C.
Supplier:  LabScientific XH2002
Encompass
Slide Warmer with Temperature Control LED 120V; 110V. 10" x 7". Surface holds about 23 slides. Shipping weight: 10lbs. Each of our slide warmers come with easy to operate push-button control panels with modern LED screens to view temperature in real time. The anodized aluminum surface provides heat control across the unit. The dark color of the surface also provides contrast against the samples. Each unit can reach up to 167 degrees Fahrenheit.
Catalog No. NC0963586