Learn More
Fire King Security Group Fire Resistant File Cabinet - 2 Drawer Let/Leg. 22" depth Parchment Finish
SDP
Supplier:  Fire King Security Group 2R1822C
Encompass_Preferred
Two-drawer vertical letter / legal-size 22" depth UL Class 350 one-hour fire / impact rated filing cabinet with one keylock in top drawer to control all drawers. ETL Verified Mark 1/2 hour fire protection hard drives, CDs, DVDs, flash drives.
Catalog No. NC1776930