Learn More
Genetex Inc SMARCA2 Monoclonal Antibody [HMV337] HistoMAX™ (GTX640917) GeneTex - IHC-Optimized Antibody
SDP
Supplier:  Genetex Inc GTX640917
Encompass_Preferred
The SMARCA2 antibody [HMV337] HistoMAX™ (GTX640917) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human warthin tumor (WT) from salivary gland tissue using GTX640917 SMARCA2 antibody [HMV337] HistoMAX. Warthin tumor? showing a strong SMARCA2 immunostaining of tumor cells.
Catalog No. NC3669370