Learn More
Aviva Systems Biology Corp SMARCA3 Antibody - N-terminal region (ARP34514_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34514_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SMARCA3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SMARCA3. Alternate names: ZBU1, HLTF1, RNF80, HIP116, SNF2L3, HIP116A, SMARCA3ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55432