Learn More
Aviva Systems Biology Corp SMARCD3 Antibody - N-terminal region (ARP35652_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35652_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMARCD3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SMARCD3. Alternate names: BAF60C, CRACD3, MGC111010, Rsc6pApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55806