Learn More
Trajan Scientific Smart SPME Fib 85Um Grey 5 Pcs
Supplier:  Trajan Scientific SFIBA85/10P5
Encompass
Smart SPME fiber Acrylate (Polyacrylate) Fiber thickness 85µm Fiber length 10mm color code: grey pack of 5pcs
Catalog No. 501920860