Learn More
Perkin Elmer US LLC SMARTintro PFA-Platinum HF-Resistant Sample Introduction Kit
Supplier:  Perkin Elmer US LLC N8150033
Encompass_Preferred
PerkinElmer introduces the NexION 2000 ICP-MS PFA-platinum solution-ready sample introduction technology. Engineered for analysis in the semiconductor industry, this high-performance HF-resistant system utilizes a PFA nebulizer, platinum injector, and a demountable SilQ ultra high purity quartz torch to minimize contamination and deliver absolute accuracy and precision. This kit includes: PFA Barrel Spray Chamber with Additional Gas Port (N8150450) Torch Light Shield Disk (N8152377) PFA-200 Self-Aspirating Nebulizer with Ultem Probe (N8152420) 2.0 mm I.D. Demountable Platinum Injector (N8152429) Platinum Torch Cassette (N8152434) Torch Injector Nut Removal Tool (N8152443) Demountable SilQ Torch (N8152448) Gas Line (N8152449) Torch Injector Nut (N8152451) 0.8 mm I.D. PTFE Drain Line Tubing with 1/4-28 Fitting (N8152456) Injector Chamber Adapter (N8152551)
Catalog No. 50-211-1322
Only null left
Add to Cart
Request a Quote