Learn More
Medchemexpress LLC HY-101070 10mg , SMCC-DM1 CAS:1228105-51-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10107010MG
Encompass_Preferred
HY-101070 10mg SMCC-DM1 CAS: 1228105-51-8 SMCC-DM1 (DM1-SMCC) is a drug-linker conjugate composed of a potent microtubule-disrupting agent DM1 and a linker SMCC to make antibody drug conjugate (ADC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9215