Learn More
CPC Scientific H-Ser-Pro-Ser-Val-Asp-Lys-Ala-Arg-Ala-Glu-Leu-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific HCAG001B
Encompass
One of the N-Y antigen epitopes has been identified as the 11 amino acid peptide SPSVDKARAEL derived from SMCY, an evolutionarily conserved protein encoded on the Y chromosome. T2-B7 cells incubated with this peptide were lysed by the H-Y specific CTL clone. Half maximal lysis was achieved at 10 pMONE-LETTER SEQUENCE: SPSVDKARAELMOLECULAR FORMULA: C49H85N15O18MOLECULAR WEIGHT:1172.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: ImmunomodulationREFERENCES: W.Wang et al., Science, 269, 1588 (1995)
Catalog No. 50-210-9413