Learn More
ABclonal Technology SMG7 Rabbit pAb
SDP
Supplier:  ABclonal Technology A153711000UL
Encompass_Preferred
This gene encodes a protein that is essential for nonsense-mediated mRNA decay (NMD), a process whereby transcripts with premature termination codons are targeted for rapid degradation by a mRNA decay complex. The mRNA decay complex consists, in part, of this protein along with proteins SMG5 and UPF1. The N-terminal domain of this protein is thought to mediate its association with SMG5 or UPF1 while the C-terminal domain interacts with the mRNA decay complex. This protein may therefore couple changes in UPF1 phosphorylation state to the degradation of NMD-candidate transcripts. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Catalog No. 50-001-07719