Learn More
Enzo Life Sciences SMI-4a (5mg). CAS: 327033-36-3
SDP
Supplier:  Enzo Life Sciences ALX270515M005
Encompass_Preferred
Selective inhibitor of Pim-1 and Pim-2 protein kinases. Inducer of G1 phase cell cycle arrest. Inducer of p27Kip1. Inducer of apoptosis through the mitocondrial pathway. Inhibitor of the mammalian target of rapamycin C1 (mTORC1) pathway. Downregulates c-myc. Inhibitor of PRAS40 phosphorylation and mTOR activity. Potential anti-cancer compound. Blocks prostate cancer growth. Purity: ≥95% (HPLC, TLC, NMR/ELSD). Solubility: Soluble in DMSO (at least 5mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8632