Learn More
Abcam SMI-4a, ATP-competitive Pim kinase inhibitor, 25MG
Supplier:  Abcam AB14136725MG
Encompass_Preferred
MW 273.23 Da. Potent, selective, cell-permeable, ATP-competitive Pim-1 kinase inhibitor (IC₅₀ = 17 nM). Moderately potent against Pim-2 kinase. Selective versus a panel of other serine/threonine and tyrosine kinases. Inhibits the growth of myeloid and lymphoid cell lines. Induces cell cycle arrest in leukemia and prostate cancer cells.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6662