Learn More
Medchemexpress LLC Smifh2 | 340316-62-3 | 99.1% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1693110MG
Encompass_Preferred
SMIFH2 is a small-molecule formin inhibitor used in research to inhibit actin polymerization and perturb the actin cytoskeleton. It is supplied as a high-purity solid for laboratory use and is intended for research use only.
  • Inhibits formin-mediated actin polymerization in vitro.
  • Enables study of cytoskeletal dynamics and formin function.
  • Supplied as a high-purity solid suitable for biochemical assays.
  • Available in small-mass vials convenient for lab experiments.
  • Soluble in DMSO for preparation of concentrated stock solutions.
Catalog No. 50-002-98497