Learn More
Medchemexpress LLC HY-16931 5mg , SMIFH2 CAS:340316-62-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY169315MG
Encompass_Preferred
HY-16931 5mg SMIFH2 CAS: 340316-62-3 SMIFH2 is a formin specific inhibitor. SMIFH2 inhibits actin polymerization by Formins and affects the actin cytoskeleton. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0198