Learn More
Selleck Chemical LLC Smilagenin E5001-5MG
SDP
Supplier:  Selleck Chemical LLC E50015MG
Encompass_Preferred
Smilagenin (SMI) is a small-molecule steroidal sapogenin from Anemarrhena asphodeloides and Pelargonium hortorum widely used in traditional Chinese medicine for treating chronic neurodegeneration diseases Smilagenin (SMI) improves memory of aged rats by increasing the muscarinic receptor subtype 1 (M1)-receptor density Smilagenin (SMI) attenuates A (25-35)-induced neurodegenerationvia stimulating the gene expression of brain-derived neurotrophic factor may represents a novel therapeutic strategy for AD
Catalog No. 50-000-41178