Learn More
Medchemexpress LLC Smip34 | 1189710-20-0 | 90.0% | 10 MG

Supplier: Medchemexpress LLC HY16990310MG
SMIP34 is a small-molecule PELP1 inhibitor used in cancer research to inhibit cell proliferation and tumor progression. It is provided as a white to off-white solid in small research quantities, suitable for in vitro studies of breast and endometrial cancer.
- PELP1 inhibitor that reduces cell viability and colony formation.
- Binds PELP1 with a Kd of 37.4 μM.
- CAS number 1189710-20-0 for unambiguous identification.
- Molecular formula C25H32ClN5O3 and molecular weight 486.01 g/mol.
- Purity approximately 90.0% as supplied.
- White to off-white solid suitable for research use.
- Store at -20°C, protect from light, and handle under inert atmosphere when required.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.