Learn More
New England Biolabs, Inc. SmlI – 2500 units
SDP
Supplier:  New England Biolabs, Inc. R0597L
Encompass
A restriction endonuclease that recognizes the sequence C^TYRA_G.
Catalog No. 50-812-315