Learn More
Selleck Chemical LLC Smoothened Agonist (SAG) HCl 5mg 2095432-58-7
SDP
Supplier:  Selleck Chemical LLC S77795MG
Encompass_Preferred
Smoothened Agonist (SAG) HCl is a cell-permeable Smoothened (Smo) agonist with EC50 of 3 nM in Shh-LIGHT2 cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6382