Learn More
Medchemexpress LLC SMPDL3A Mouse HEK2 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7720950UG
Encompass_Preferred
SMPDL3A protein hydrolyzes nucleotide triphosphates particularly ATP and p-nitrophenyl-TMP It has limited activity with nucleoside diphosphates and none with nucleoside monophosphates SMPDL3A also catalyzes the conversion of CDP-choline to CMP and phosphocholine and CDP-ethanolamine However it lacks sphingomyelin phosphodiesterase activity SMPDL3A Protein Mouse (HEK293 His) is the recombinant mouse-derived SMPDL3A protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-86509