Learn More
Novus Biologicals Simple Plex Human CXCL10/IP-10 Assay Kit, 1 Kit (1 Analyte x 72 Samples) /
SDP
Supplier:  Novus Biologicals ST01BPS000237
Encompass
Contains cartridge, sample diluent SD13, and wash buffer for running samples on ELLA (Ella - Automated Immunoassay Platform | ProteinSimple). Sample type indicated with PS (Plasma/Serum).
Catalog No. NC2737904