Learn More
Medchemexpress LLC Smpt | 112241-19-7 | 97.4% | 388.46 g·mol⁻¹ | C18H16N2O4S2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY126405100MG
Encompass_Preferred
SMPT is a non-cleavable linker used to connect cytotoxic payloads to antibodies during antibody-drug conjugate (ADC) synthesis. It is supplied as a light yellow solid with formula C18H16N2O4S2, molecular weight 388.46 g·mol⁻¹, and a typical purity of 97.4%, intended for research use and available in multiple package sizes.
  • Non-cleavable linker suitable for stable ADC constructs.
  • High purity (97.4%) for reproducible conjugation.
  • Light yellow solid that is easy to weigh and handle.
  • Defined molecular weight and formula for analytical consistency.
  • Available in multiple package sizes to support different research scales.
Catalog No. 50-002-02951