Learn More
MSC KleenGuard Disposable Lab Coat, SMS, White, 2XL, 25/Pk.
Supplier:  MSC 66314899
Encompass
These disposable lab coats are designed for cleanroom environments, offering protection and durability. The SMS fabric construction provides a barrier against particles, making them suitable for controlled settings.
  • Features a white color and 2XL size
  • Equipped with a snap-front closure for easy donning and removal
  • Designed with an open wrist style for comfort
  • Includes two pockets for convenient storage of small items
  • Disposable design for ease of use and maintenance
Catalog No. 50-006-87698