Learn More
Medchemexpress LLC HY-112713 5mg Medchemexpress, SMS2-IN-2 CAS:2241838-28-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1127135MG
Encompass_Preferred
Medchemexpress, HY-112713 5mg SMS2-IN-2 CAS:2241838-28-6 SMS2-IN-2 is a potent, highly selective and orally active sphingomyelin synthase 2 (SMS2) inhibitor, with IC50s of 100 nM and 56 μM for SMS2 and SMS1, respectively. Anti-chronic inflammatory activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9152