Learn More
Medchemexpress LLC HY-145010 5mg , SN-011 CAS:2249435-90-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1450105MG
Encompass_Preferred
HY-145010 5mg SN-011 CAS: 2249435-90-1 SN-011 is a potent and selective mouse and human STING inhibitor, with an IC50 of 76 nM for STING signaling. SN-011 competes with cyclic dinucleotide (CDN) for the binding pocket of the STING dimer, blocking CDN binding and STING activation. SN-011 can be used for the research of STING-driven autoimmune and inflammatory disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9519