Learn More
Echelon Biosciences Research Labs SN-1-BIOTIN MONOLYSOCARDIOLIPIN 10 ug
SDP
Supplier:  Echelon Biosciences Research Labs LMB1610UG
Encompass
sn-1-Biotin-Labeled Monolysocardiolipin (Biotin-monolysocardiolipin) can be used in microplate and bead-based assays and affinity experiments. Cardiolipin (CL) is an important component inprokaryotic and in eukaryotic inner mitochondrial membranes. In humans, CL is made by cardiolipin synthase from phosphatidylglycerol and cytidinediphosphate-diacylglycerol. Monolysocardiolipin is an intermediate in CL remodeling, and acetyl transferase tafazzin mutations impairing linoleic acid transfer to MCL are considered the main reason of Barth syndrome. Autoantibodies to CL have been found in patients with anti-phospholipid syndrome and lupus. This item is non returnable
Catalog No. 50-822-27