Learn More
Medchemexpress LLC Sn-glycero-3-phosphocholine | 28319-77-9 | ≥98.0% | 257.22 | 50 G
SDP
Supplier:  Medchemexpress LLC HY1755250G
Encompass_Preferred
sn-Glycero-3-phosphocholine is a precursor in the biosynthesis of brain phospholipids and increases the bioavailability of choline in nervous tissue. It has significant effects on cognitive function with a good safety profile and tolerability, and is effective in the treatment of Alzheimer's disease and dementia.
  • Cholinesterase (ChE)
  • Endogenous metabolite
  • Neurological disease
  • Natural product
  • Microorganism
  • Source classification
Catalog No. 50-003-86271